CAS RN 632-69-9
CAS RN 632-69-9
Thermo Scientific Chemicals Rose Bengal, 85%, pure, high purity, Biological stain
CAS: 632-69-9 Molecular Formula: C20H4Cl4I4Na2O5 Molecular Weight (g/mol): 1019.644 MDL Number: MFCD00005043 InChI Key: DPBQSRZQYAYDGY-UHFFFAOYSA-N Synonym: Acid Red 94,C.I. 45440 PubChem CID: 87244310 IUPAC Name: sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one SMILES: C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]
Thermo Scientific Chemicals Rose Bengal, 85%, pure, certified
CAS: 632-69-9 Molecular Formula: C20H4Cl4I4Na2O5 Molecular Weight (g/mol): 1019.644 MDL Number: MFCD00005043 InChI Key: DPBQSRZQYAYDGY-UHFFFAOYSA-N Synonym: Acid Red 94,C.I. 45440 PubChem CID: 87244310 IUPAC Name: sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one SMILES: C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]
Thermo Scientific Chemicals Rose Bengal
CAS: 632-69-9 Molecular Formula: C20H4Cl4I4Na2O5 Molecular Weight (g/mol): 1019.644 MDL Number: MFCD00151169 InChI Key: DPBQSRZQYAYDGY-UHFFFAOYSA-N Synonym: rose bengal disodium salt,r105 sodium,rose bengal sodium,rose-bengal 131 i natrium,food red no. 105, sodium salt,food red color no. 105, sodium salt,rosa bengala sodica 131 i inn-spanish,rose bengale 131 i sodique inn-french,sel disodique de rose bengale iodee 131 i,roseum bengalense 131 i natricum inn-latin PubChem CID: 87244310 IUPAC Name: sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one SMILES: C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]
Rose Bengal, Sigma-Aldrich
CAS: 632-69-9
Rose Bengal, Certified, Sigma-Aldrich
CAS: 632-69-9 Molecular Formula: C20H2Cl4I4Na2O5 Molecular Weight (g/mol): 1017.63 InChI Key: KCQREHTWEUECQT-UHFFFAOYSA-L IUPAC Name: disodium 4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate) SMILES: [Na+].[Na+].[O-]C1=C(I)C=C2C(OC3=C(I)C([O-])=C(I)C=C3C22OC(=O)C3=C(Cl)C(Cl)=C(Cl)C(Cl)=C23)=C1I
Rose Bengal, Spectrum™ Chemical
CAS: 632-69-9
Food Red No. 105, TCI America™
CAS: 632-69-9 Molecular Formula: C20H4Cl4I4Na2O5 Molecular Weight (g/mol): 1019.644 MDL Number: MFCD00151169 InChI Key: DPBQSRZQYAYDGY-UHFFFAOYSA-N PubChem CID: 87244310 IUPAC Name: sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one SMILES: C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]
Acid Red 94, TCI America™
CAS: 632-69-9 Molecular Formula: C20H4Cl4I4Na2O5 Molecular Weight (g/mol): 1019.644 MDL Number: MFCD00151169 InChI Key: DPBQSRZQYAYDGY-UHFFFAOYSA-N Synonym: Rose Bengal PubChem CID: 87244310 IUPAC Name: sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one SMILES: C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]
Fisher Science Education™ Rose Bengal Solution
CAS: 632-69-9 Molecular Formula: C20H4Cl4I4Na2O5 Molecular Weight (g/mol): 1019.644 InChI Key: DPBQSRZQYAYDGY-UHFFFAOYSA-N Synonym: Acid Red 94,C.I. 45440 PubChem CID: 87244310 IUPAC Name: sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one SMILES: C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]
Rose Bengal Disodium Salt (Certified Biological Stain), Fisher Chemical
CAS: 632-69-9 Molecular Formula: C20H4Cl4I4Na2O5 Molecular Weight (g/mol): 1019.644 MDL Number: MFCD00151169 InChI Key: DPBQSRZQYAYDGY-UHFFFAOYSA-N Synonym: Acid Red 94 PubChem CID: 87244310 IUPAC Name: sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one SMILES: C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]