Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Food Red No. 105, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 g
Specifications
Specifications
| Formula Weight | 1017.63 |
| Color | Red |
| Quantity | 25 g |
| Physical Form | Powder |
| Chemical Name or Material | Food Red No. 105 |
Safety and Handling
| EINECSNumber | (4)-0650 |
| RTECSNumber | LM5920000 |
| TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction