accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94744-50-0

H3CCH3OOHHNOO

CAS RN 94744-50-0

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methylpropanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2-methylpropanoic acid
Molecular Weight (g/mol): 325.36
Molecular Formula: C19H19NO4
InChi Key: HOZZVEPRYYCBTO-UHFFFAOYSA-N
SMILES: CC(C)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Grade 
  • (4)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)