accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 943-29-3

CH3H3CCH3OHO

CAS RN 943-29-3

IUPAC Name: 4-tert-butylcyclohexane-1-carboxylic acid
Synonyms: 4-tert-butylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: QVQKEGYITJBHRQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1CCC(CC1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results