accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 928-92-7

CH3OH(E)

CAS RN 928-92-7

IUPAC Name: (4E)-hex-4-en-1-ol
Synonyms: (4E)-hex-4-en-1-ol
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: VTIODUHBZHNXFP-NSCUHMNNSA-N
SMILES: C\C=C\CCCO
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results