accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 927-68-4

H3COOBr

CAS RN 927-68-4

IUPAC Name: 2-bromoethyl acetate
Synonyms: ethanol, 2-bromo-, acetate
Molecular Weight (g/mol): 167
Molecular Formula: C4H7BrO2
InChi Key: RGHQKFQZGLKBCF-UHFFFAOYSA-N
SMILES: CC(=O)OCCBr
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results