accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 920519-32-0

OOHOOOOOONNH(S)(R)CH3CH3H3C(S)CH3CH3H3C

CAS RN 920519-32-0

IUPAC Name: (4S,5R)-3-[(2S)-4-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoyl]-2,2,5-trimethyl-1,3-oxazolidine-4-carboxylic acid
Synonyms: (4S,5R)-3-[(2S)-4-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-oxobutanoyl]-2,2,5-trimethyl-1,3-oxazolidine-4-carboxylic acid
Molecular Weight (g/mol): 552.62
Molecular Formula: C30H36N2O8
InChi Key: NKQJRYSIZOXCTJ-LAMXGVLKSA-N
SMILES: C[C@H]1OC(C)(C)N([C@@H]1C(O)=O)C(=O)[C@H](CC(=O)OC(C)(C)C)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)