accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 81028-91-3

H2OH2OH2OH2OH2OH2OH2OH2ONaNaNaHO(S)(S)HOHOOPOOOO(R)OPOOO

CAS RN 81028-91-3

IUPAC Name: trisodium octahydrate [(3S,4S,5R)-2,3,4-trihydroxy-5-[(phosphonatooxy)methyl]oxolan-2-yl]methyl phosphate
Synonyms: trisodium fructose diphosphate
Molecular Weight (g/mol): 549.17
Molecular Formula: C6H26Na3O20P2
InChi Key: DBTJWGHAMYSWQR-OLMHPWOBSA-J
SMILES: O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].O[C@H]1[C@H](O)C(O)(COP([O-])([O-])=O)O[C@@H]1COP([O-])([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results