accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76195-82-9

CH3H3CHNNH2CH3HCl

CAS RN 76195-82-9

IUPAC Name: (2,4,6-trimethylphenyl)hydrazine hydrochloride
Synonyms: (2,4,6-trimethylphenyl)hydrazine hydrochloride
Molecular Weight (g/mol): 186.68
Molecular Formula: C9H15ClN2
InChi Key: XFGPCULNJGZRTH-UHFFFAOYSA-N
SMILES: Cl.CC1=CC(C)=C(NN)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results