CAS RN 75308-46-2
CAS RN 75308-46-2
IUPAC Name:
tert-butyl 2,6-dichloropyridine-4-carboxylate
Synonyms:
tert-butyl 2,6-dichloropyridine-4-carboxylate
Molecular Weight (g/mol):
248.1
Molecular Formula:
C10H11Cl2NO2
InChi Key:
PLRVIRHWKQKSAK-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)C1=CC(Cl)=NC(Cl)=C1
1
–
1
of
1
results
1
tert-Butyl 2,6-Dichloroisonicotinate 98.0+%, TCI America™
CAS: 75308-46-2 Molecular Formula: C10H11Cl2NO2 Molecular Weight (g/mol): 248.10 MDL Number: MFCD08445655 InChI Key: PLRVIRHWKQKSAK-UHFFFAOYSA-N Synonym: 2,6-Dichloroisonicotinic Acid tert-Butyl Ester, tert-Butyl 2,6-Dichloro-4-pyridinecarboxylate, 2,6-Dichloro-4-pyridinecarboxylic Acid tert-Butyl Ester PubChem CID: 22594123 IUPAC Name: tert-butyl 2,6-dichloropyridine-4-carboxylate SMILES: CC(C)(C)OC(=O)C1=CC(Cl)=NC(Cl)=C1