CAS RN 74563-64-7
CAS RN 74563-64-7
IUPAC Name:
3,7,11,15-tetramethylhexadecane-1,2,3-triol
Synonyms:
phytantriol
Molecular Weight (g/mol):
330.55
Molecular Formula:
C20H42O3
InChi Key:
CGIHFIDULQUVJG-UHFFFAOYNA-N
SMILES:
CC(C)CCCC(C)CCCC(C)CCCC(C)(O)C(O)CO
1
–
1
of
1
results
1
Phytantriol (mixture of isomers) 95.0+%, TCI America™
CAS: 74563-64-7 Molecular Formula: C20H42O3 Molecular Weight (g/mol): 330.55 MDL Number: MFCD00129085 InChI Key: CGIHFIDULQUVJG-UHFFFAOYNA-N Synonym: 3,7,11,15-Tetramethylhexadecane-1,2,3-triol, 1,2,3-Trihydroxy-3,7,11,15-tetramethylhexadecane PubChem CID: 3018525 ChEBI: CHEBI:47770 IUPAC Name: 3,7,11,15-tetramethylhexadecane-1,2,3-triol SMILES: CC(C)CCCC(C)CCCC(C)CCCC(C)(O)C(O)CO