accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73870-24-3

HBrBrN

CAS RN 73870-24-3

IUPAC Name: hydrogen 4-(bromomethyl)pyridine bromide
Synonyms: hydrogen 4-(bromomethyl)pyridine bromide
Molecular Weight (g/mol): 252.94
Molecular Formula: C6H7Br2N
InChi Key: VAJUUDUWDNCECT-UHFFFAOYSA-N
SMILES: [H+].[Br-].BrCC1=CC=NC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)