accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71989-16-7

H2NONHOOOHO

CAS RN 71989-16-7

IUPAC Name: 3-carbamoyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: 3-carbamoyl-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 354.36
Molecular Formula: C19H18N2O5
InChi Key: YUGBZNJSGOBFOV-UHFFFAOYNA-N
SMILES: NC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)