accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71916-82-0

FClBr

CAS RN 71916-82-0

IUPAC Name: 1-(bromomethyl)-4-chloro-2-fluorobenzene
Synonyms: 1-(bromomethyl)-4-chloro-2-fluorobenzene
Molecular Weight (g/mol): 223.47
Molecular Formula: C7H5BrClF
InChi Key: UFCSSWZQROEFBZ-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC=C1CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results