CAS RN 7189-82-4
CAS RN 7189-82-4
IUPAC Name:
2,2'-bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-2'H-1,2'-biimidazole
Synonyms:
2,2'-bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,2'-biimidazole
Molecular Weight (g/mol):
659.61
Molecular Formula:
C42H28Cl2N4
InChi Key:
MHDULSOPQSUKBQ-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC=C1C1=NC(=C(N1C1(N=C(C(=N1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1Cl)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2,2'-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,2'-biimidazole 97.0+%, TCI America™
CAS: 7189-82-4 Molecular Formula: C42H28Cl2N4 Molecular Weight (g/mol): 659.61 MDL Number: MFCD09028023 InChI Key: MHDULSOPQSUKBQ-UHFFFAOYSA-N PubChem CID: 81610 IUPAC Name: 2,2'-bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-2'H-1,2'-biimidazole SMILES: ClC1=CC=CC=C1C1=NC(=C(N1C1(N=C(C(=N1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1Cl)C1=CC=CC=C1)C1=CC=CC=C1