accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7065-46-5

H3CH3CCH3ClO

CAS RN 7065-46-5

IUPAC Name: 3,3-dimethylbutanoyl chloride
Synonyms: 3,3-dimethylbutanoyl chloride
Molecular Weight (g/mol): 134.6
Molecular Formula: C6H11ClO
InChi Key: BUTKIHRNYUEGKB-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(Cl)=O
Boiling Point 
  • (1)
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (6)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (5)
  • (3)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)