accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 693-58-3

H3CBr

CAS RN 693-58-3

IUPAC Name: 1-bromononane
Synonyms: 1-bromo-N-nonane
Molecular Weight (g/mol): 207.16
Molecular Formula: C9H19Br
InChi Key: AYMUQTNXKPEMLM-UHFFFAOYSA-N
SMILES: CCCCCCCCCBr
Boiling Point 
  • (3)
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (6)