CAS RN 6578-06-9
CAS RN 6578-06-9
IUPAC Name:
4-methylaniline; [(5-bromo-4-chloro-1H-indol-3-yl)oxy]phosphonic acid
Synonyms:
(5-bromo-4-chloro-1H-indol-3-yl)oxyphosphonic acid; toluidine
Molecular Weight (g/mol):
433.62
Molecular Formula:
C15H15BrClN2O4P
InChi Key:
QEIFSLUFHRCVQL-UHFFFAOYSA-N
SMILES:
CC1=CC=C(N)C=C1.OP(O)(=O)OC1=CNC2=CC=C(Br)C(Cl)=C12
1
–
2
of
2
results
1
5-Bromo-4-chloro-3-indolyl Phosphate p-Toluidine Salt 98.0+%, TCI America™
CAS: 6578-06-9 Molecular Formula: C15H15BrClN2O4P Molecular Weight (g/mol): 433.62 MDL Number: MFCD00040636 InChI Key: QEIFSLUFHRCVQL-UHFFFAOYSA-N Synonym: X-Phosphate p-Toluidine Salt
2
Fisher Scientific 5-Bromo-4-chloro-3-indolyl Phosph., p-Toluidine Salt (Wht. to Off-Wht. Powd.), Fisher BioReagents
Molecular Formula: C15H15BrClN2O4P Molecular Weight (g/mol): 433.62 InChI Key: QEIFSLUFHRCVQL-UHFFFAOYSA-N