accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 632-51-9

CAS RN 632-51-9

IUPAC Name: (1,2,2-triphenylethenyl)benzene
Synonyms: tetraphenylethylene
Molecular Weight (g/mol): 332.45
Molecular Formula: C26H20
InChi Key: JLZUZNKTTIRERF-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C(=C(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)