CAS RN 63126-52-3
CAS RN 63126-52-3
IUPAC Name:
(2S,3S)-2,3-dihydroxy-N,N,N',N'-tetramethylbutanediamide
Synonyms:
(2S,3S)-2,3-dihydroxy-N,N,N',N'-tetramethylsuccinamide
Molecular Weight (g/mol):
204.23
Molecular Formula:
C8H16N2O4
InChi Key:
PCYDYHRBODKVEL-WDSKDSINSA-N
SMILES:
CN(C)C(=O)[C@@H](O)[C@H](O)C(=O)N(C)C
1
–
1
of
1
results
1
(-)-N,N,N',N'-Tetramethyl-D-tartardiamide 98.0+%, TCI America™
CAS: 63126-52-3 Molecular Formula: C8H16N2O4 Molecular Weight (g/mol): 204.23 MDL Number: MFCD00077879 InChI Key: PCYDYHRBODKVEL-WDSKDSINSA-N PubChem CID: 6999257 IUPAC Name: (2S,3S)-2,3-dihydroxy-N,N,N',N'-tetramethylbutanediamide SMILES: CN(C)C(=O)[C@@H](O)[C@H](O)C(=O)N(C)C