CAS RN 63091-82-7
CAS RN 63091-82-7
IUPAC Name:
(4S)-3-[(tert-butoxy)carbonyl]-1,3-thiazolidine-4-carboxylic acid
Synonyms:
(4S)-3-(tert-butoxycarbonyl)-1,3-thiazolidine-4-carboxylic acid
Molecular Weight (g/mol):
233.28
Molecular Formula:
C9H15NO4S
InChi Key:
FJWNZTPXVSWUKF-ZCFIWIBFSA-N
SMILES:
CC(C)(C)OC(=O)N1CSC[C@@H]1C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Boc-D-Thz-OH, Advanced ChemTech
CAS: 63091-82-7 Molecular Formula: C9H15NO4S Molecular Weight (g/mol): 233.28 MDL Number: MFCD00077001 InChI Key: FJWNZTPXVSWUKF-ZCFIWIBFSA-N IUPAC Name: (4S)-3-[(tert-butoxy)carbonyl]-1,3-thiazolidine-4-carboxylic acid SMILES: CC(C)(C)OC(=O)N1CSC[C@@H]1C(O)=O