accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 629-80-1

H3CO

CAS RN 629-80-1

IUPAC Name: hexadecanal
Synonyms: hexadecanal palmitoyl aldehyde PLM PLY N-hexadecanal
Molecular Weight (g/mol): 240.43
Molecular Formula: C16H32O
InChi Key: NIOYUNMRJMEDGI-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results