accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 624-92-0

CH3SSH3C

CAS RN 624-92-0

IUPAC Name: (methyldisulfanyl)methane
Synonyms: dimethyl disulfide DMDS disulfide dimethyl methyl disulfide (substance) sulfa-hitech methyldisulfide sulfa-hitech 0382
Molecular Weight (g/mol): 94.19
Molecular Formula: C2H6S2
InChi Key: WQOXQRCZOLPYPM-UHFFFAOYSA-N
SMILES: CSSC
Molecular Weight (g/mol) 
  • (8)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (7)
Boiling Point 
  • (2)
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (7)
  • (1)
  • (8)
  • (3)
  • (1)
  • (3)
  • (7)
  • (2)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)