CAS RN 607-34-1
CAS RN 607-34-1
IUPAC Name:
5-nitroquinoline
Synonyms:
5-nitroquinoline
Molecular Weight (g/mol):
174.16
Molecular Formula:
C9H6N2O2
InChi Key:
NDDZXHOCOKCNBM-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=C2C=CC=NC2=CC=C1
1
–
1
of
1
results
1
5-Nitroquinoline 98.0+%, TCI America™
CAS: 607-34-1 Molecular Formula: C9H6N2O2 Molecular Weight (g/mol): 174.159 MDL Number: MFCD00006790 InChI Key: NDDZXHOCOKCNBM-UHFFFAOYSA-N Synonym: quinoline, 5-nitro,5-nitro-quinoline,zlchem 399,pubchem5821,acmc-209mlr,5-nitroquinoline,5-20-07-00325 beilstein handbook reference,ksc355k8h,7ze PubChem CID: 11829 IUPAC Name: 5-nitroquinoline SMILES: C1=CC2=C(C=CC=N2)C(=C1)[N+](=O)[O-]