accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 601-75-2

H3COHOOHO

CAS RN 601-75-2

IUPAC Name: 2-ethylpropanedioic acid
Synonyms: ethylmalonic acid ethylmalonate (substance)
Molecular Weight (g/mol): 132.12
Molecular Formula: C5H8O4
InChi Key: UKFXDFUAPNAMPJ-UHFFFAOYSA-N
SMILES: CCC(C(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)