CAS RN 60-81-1
CAS RN 60-81-1
IUPAC Name:
1-(2,4-dihydroxy-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}phenyl)-3-(4-hydroxyphenyl)propan-1-one
Synonyms:
phlorizin phloridzin phloretin 2'-glucoside phlorhizin phlorizoside floridzin phlorrhizin
Molecular Weight (g/mol):
436.41
Molecular Formula:
C21H24O10
InChi Key:
IOUVKUPGCMBWBT-QNDFHXLGSA-N
SMILES:
OC[C@H]1O[C@@H](OC2=CC(O)=CC(O)=C2C(=O)CCC2=CC=C(O)C=C2)[C@H](O)[C@@H](O)[C@@H]1O
1
–
1
of
1
results
1
Phlorizin Hydrate 97.0+%, TCI America™
CAS: 60-81-1 Molecular Formula: C21H24O10 Molecular Weight (g/mol): 436.413 MDL Number: MFCD00006591 InChI Key: IOUVKUPGCMBWBT-QNDFHXLGSA-N Synonym: phlorizin,phloridzin,phlorhizin,phlorizoside,floridzin,phlorrhizin,phloretin 2'-glucoside,phloridzosid,phlorrhizen,phlorizine PubChem CID: 6072 ChEBI: CHEBI:8113 IUPAC Name: 1-[2,4-dihydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]-3-(4-hydroxyphenyl)propan-1-one SMILES: C1=CC(=CC=C1CCC(=O)C2=C(C=C(C=C2OC3C(C(C(C(O3)CO)O)O)O)O)O)O