accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5968-90-1

OH(R)O(R)(R)OH(S)OHNNONHONH

CAS RN 5968-90-1

IUPAC Name: 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2,3,6,9-tetrahydro-1H-purine-2,6-dione
Synonyms: xanthosine xanthine riboside 9-delta-ribofuranosylxanthine β-D-ribofuranoside, xanthine-9 xanthine 9-β-D-ribofuranoside xanthine-9b-D-ribofuranoside
Molecular Weight (g/mol): 284.23
Molecular Formula: C10H12N4O6
InChi Key: UBORTCNDUKBEOP-UUOKFMHZSA-N
SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1NC(=O)NC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results