CAS RN 5720-06-9
CAS RN 5720-06-9
2-Methoxyphenylboronic acid, 97%
CAS: 5720-06-9 Molecular Formula: C7H9BO3 Molecular Weight (g/mol): 151.96 MDL Number: MFCD00236047 InChI Key: ROEQGIFOWRQYHD-UHFFFAOYSA-N SMILES: COC1=CC=CC=C1B(O)O
2-Methoxybenzeneboronic acid, 97%
CAS: 5720-06-9 Molecular Formula: C7H9BO3 Molecular Weight (g/mol): 151.96 MDL Number: MFCD00236047 InChI Key: ROEQGIFOWRQYHD-UHFFFAOYSA-N Synonym: 2-methoxybenzeneboronic acid,2-methoxyphenyl boronic acid,o-methoxyphenylboronic acid,2-methoxyphenyl boranediol,2-boronoanisole,2-methoxypenylboronic acid,o-methoxyphenyl boronic acid,2-methoxy phenylboronic acid,2-methoxybenzene boronic acid PubChem CID: 2733958 IUPAC Name: (2-methoxyphenyl)boronic acid SMILES: COC1=CC=CC=C1B(O)O
2-Methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Molecular Formula: C7H9BO3 Molecular Weight (g/mol): 151.96 MDL Number: MFCD00236047 InChI Key: ROEQGIFOWRQYHD-UHFFFAOYSA-N PubChem CID: 2733958 IUPAC Name: (2-methoxyphenyl)boronic acid