CAS RN 548-24-3
CAS RN 548-24-3
IUPAC Name:
disodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Synonyms:
eosin B
Molecular Weight (g/mol):
624.06
Molecular Formula:
C20H6Br2N2Na2O9
InChi Key:
GYYTYUGGVBYJHE-UHFFFAOYSA-L
SMILES:
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(C=C12)[N+]([O-])=O)[N+]([O-])=O
1
–
4
of
4
results
1
Eosin B, pure, certified
CAS: 548-24-3 Molecular Formula: C20H6Br2N2Na2O9 MDL Number: MFCD00005041 Synonym: 4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt,4', 5'-Dibromo-2', 7'-dinitrofluorescein,Eosin Bluish
2
Eosin B
CAS: 548-24-3 Molecular Formula: C20H6Br2N2Na2O9 MDL Number: MFCD00005041 Synonym: C.I. 45400; 4,5-Dibromo-2,7-dinitrofluorescein disodium salt
3
Thermo Scientific Chemicals Eosin B, pure, high purity, biological stain
CAS: 548-24-3 Molecular Formula: C20H6Br2N2Na2O9 Molecular Weight (g/mol): 624.06 MDL Number: MFCD00005041 Synonym: 4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt,4', 5'-Dibromo-2', 7'-dinitrofluorescein,Eosin Bluish
4
Acid Red 91, TCI America™
CAS: 548-24-3 Molecular Formula: C20H8Br2N2O9 MDL Number: MFCD00005041 Synonym: Eosin Bluish, Imperial Red