accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54296-25-2

CH3N

CAS RN 54296-25-2

IUPAC Name: 4'-(4-propylphenyl)-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-(4-propylphenyl)-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 297.4
Molecular Formula: C22H19N
InChi Key: PWZKBBQJWUEPOK-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results