CAS RN 54197-66-9
CAS RN 54197-66-9
IUPAC Name:
6-hydroxy-1,2,3,4-tetrahydroquinolin-2-one
Synonyms:
6-hydroxy-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol):
163.18
Molecular Formula:
C9H9NO2
InChi Key:
HOSGXJWQVBHGLT-UHFFFAOYSA-N
SMILES:
OC1=CC=C2NC(=O)CCC2=C1
1
–
1
of
1
results
1
3,4-Dihydro-6-hydroxy-2(1H)-quinolinone 98.0+%, TCI America™
CAS: 54197-66-9 Molecular Formula: C9H9NO2 Molecular Weight (g/mol): 163.176 MDL Number: MFCD02179410 InChI Key: HOSGXJWQVBHGLT-UHFFFAOYSA-N Synonym: 6-Hydroxy-3,4-dihydro-2(1H)-quinolinone, 6-Hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline PubChem CID: 2774040 IUPAC Name: 6-hydroxy-3,4-dihydro-1H-quinolin-2-one SMILES: C1CC(=O)NC2=C1C=C(C=C2)O