accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5417-45-8

ClCH3H2NH3C

CAS RN 5417-45-8

IUPAC Name: 2,3-dimethylaniline hydrochloridyl
Synonyms: xylidine hydrochloridyl
Molecular Weight (g/mol): 156.63
Molecular Formula: C8H11ClN
InChi Key: LRUSTAAVBGFTBM-UHFFFAOYSA-N
SMILES: [Cl].CC1=CC=CC(N)=C1C
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)