CAS RN 5345-01-7
CAS RN 5345-01-7
IUPAC Name:
3-ethyl-3-methylpentanedioic acid
Synonyms:
3-ethyl-3-methylpentanedioic acid
Molecular Weight (g/mol):
174.2
Molecular Formula:
C8H14O4
InChi Key:
XAWFHZMTJUGGEE-UHFFFAOYSA-N
SMILES:
CCC(C)(CC(O)=O)CC(O)=O
1
–
1
of
1
results
1
3-Ethyl-3-methylglutaric Acid 98.0+%, TCI America™
CAS: 5345-01-7 Molecular Formula: C8H14O4 Molecular Weight (g/mol): 174.20 MDL Number: MFCD00020494 InChI Key: XAWFHZMTJUGGEE-UHFFFAOYSA-N PubChem CID: 79290 IUPAC Name: 3-ethyl-3-methylpentanedioic acid SMILES: CCC(C)(CC(=O)O)CC(=O)O