CAS RN 4532-96-1
CAS RN 4532-96-1
IUPAC Name:
1-(propan-2-yl)-1H-imidazole
Synonyms:
1-isopropylimidazole
Molecular Weight (g/mol):
110.16
Molecular Formula:
C6H10N2
InChi Key:
IPIORGCOGQZEHO-UHFFFAOYSA-N
SMILES:
CC(C)N1C=CN=C1
1
–
1
of
1
results
1
1-Isopropylimidazole 97.0+%, TCI America™
CAS: 4532-96-1 Molecular Formula: C6H10N2 Molecular Weight (g/mol): 110.16 MDL Number: MFCD00234239 InChI Key: IPIORGCOGQZEHO-UHFFFAOYSA-N PubChem CID: 10176113 IUPAC Name: 1-propan-2-ylimidazole SMILES: CC(C)N1C=CN=C1