accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42872-74-2

CH3NBr

CAS RN 42872-74-2

IUPAC Name: 3-bromo-4-methylbenzonitrile
Synonyms: 3-bromo-4-methylbenzonitrile
Molecular Weight (g/mol): 196.05
Molecular Formula: C8H6BrN
InChi Key: VXUMRYMTYKDWMO-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results