accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3739-38-6

OOO

CAS RN 3739-38-6

IUPAC Name: 3-phenoxybenzoate
Synonyms: 3-phenoxybenzoate
Molecular Weight (g/mol): 213.21
Molecular Formula: C13H9O3
InChi Key: NXTDJHZGHOFSQG-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC(OC2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results