accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 368-48-9

FFFN

CAS RN 368-48-9

IUPAC Name: 2-(trifluoromethyl)pyridine
Synonyms: 2-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 147.1
Molecular Formula: C6H4F3N
InChi Key: ATRQECRSCHYSNP-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=N1
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)