CAS RN 35274-05-6
CAS RN 35274-05-6
IUPAC Name:
hexadecyl 2-hydroxypropanoate
Synonyms:
cetyl lactate
Molecular Weight (g/mol):
314.51
Molecular Formula:
C19H38O3
InChi Key:
WUKXMJCZWYUIRZ-UHFFFAOYNA-N
SMILES:
CCCCCCCCCCCCCCCCOC(=O)C(C)O
1
–
2
of
2
results
1
Hexadecyl Lactate 75.0+%, TCI America™
CAS: 35274-05-6 Molecular Formula: C19H38O3 Molecular Weight (g/mol): 314.51 InChI Key: WUKXMJCZWYUIRZ-UHFFFAOYSA-N Synonym: Cetyl Lactate, Hexadecyl 2-Hydroxypropionate, Lactic Acid Hexadecyl Ester PubChem CID: 37098 IUPAC Name: hexadecyl 2-hydroxypropanoate SMILES: CCCCCCCCCCCCCCCCOC(=O)C(C)O
2
Sigma Aldrich hexadecyl lactate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.