CAS RN 3344-77-2
CAS RN 3344-77-2
IUPAC Name:
12-bromododecan-1-ol
Synonyms:
bromo-dodecanol C12H25BrO
Molecular Weight (g/mol):
265.24
Molecular Formula:
C12H25BrO
InChi Key:
ASIDMJNTHJYVQJ-UHFFFAOYSA-N
SMILES:
OCCCCCCCCCCCCBr
1
–
2
of
2
results
1
12-Bromo-1-dodecanol 98.0+%, TCI America™
CAS: 3344-77-2 Molecular Formula: C12H25BrO Molecular Weight (g/mol): 265.24 MDL Number: MFCD00004754 InChI Key: ASIDMJNTHJYVQJ-UHFFFAOYSA-N PubChem CID: 137895 IUPAC Name: 12-bromododecan-1-ol SMILES: OCCCCCCCCCCCCBr
2
Sigma Aldrich 12-Bromo-1-dodecanol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.