accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 320-94-5

OONOOFFF

CAS RN 320-94-5

IUPAC Name: 2-nitro-4-(trifluoromethyl)benzoate
Synonyms: 2-nitro-4-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 234.11
Molecular Formula: C8H3F3NO4
InChi Key: MYSAXQPTXWKDPQ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(C=C1[N+]([O-])=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)