CAS RN 27821-45-0
CAS RN 27821-45-0
IUPAC Name:
disodium 1-[(2R,5R)-3,4-dihydroxy-5-({[hydroxy(phosphonatooxy)phosphoryl]oxy}methyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms:
disodium 1-[(2R,5R)-3,4-dihydroxy-5-({[hydroxy(phosphonatooxy)phosphoryl]oxy}methyl)oxolan-2-yl]-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol):
448.12
Molecular Formula:
C9H12N2Na2O12P2
InChi Key:
ZQKVPFKBNNAXCE-HVQNCJTOSA-L
SMILES:
[Na+].[Na+].OC1C(O)[C@@H](O[C@@H]1COP(O)(=O)OP([O-])([O-])=O)N1C=CC(=O)NC1=O
1
–
2
of
2
results
1
Uridine-5'-diphosphate disodium salt, 98%
CAS: 27821-45-0 Molecular Formula: C9H12N2Na2O12P2 Molecular Weight (g/mol): 448.12 MDL Number: MFCD00006524,MFCD00084679,MFCD04975448 InChI Key: ZQKVPFKBNNAXCE-HVQNCJTOSA-L Synonym: 5'-udp,2na PubChem CID: 131676107 SMILES: [Na+].[Na+].OC1C(O)[C@@H](O[C@@H]1COP(O)(=O)OP([O-])([O-])=O)N1C=CC(=O)NC1=O
2
Tocris Bioscience™ UDP disodium salt
CAS: 27821-45-0 Molecular Formula: C9H12N2Na2O12P2