accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2731-73-9

NH2(R)ClOHO

CAS RN 2731-73-9

IUPAC Name: (2R)-2-amino-3-chloropropanoic acid
Synonyms: 3-chloro-L-alanine
Molecular Weight (g/mol): 123.54
Molecular Formula: C3H6ClNO2
InChi Key: ASBJGPTTYPEMLP-REOHCLBHSA-N
SMILES: N[C@@H](CCl)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results