CAS RN 2357-52-0
CAS RN 2357-52-0
IUPAC Name:
4-bromo-2-fluoro-1-methoxybenzene
Synonyms:
4-bromo-2-fluoro-1-methoxybenzene
Molecular Weight (g/mol):
205.03
Molecular Formula:
C7H6BrFO
InChi Key:
DWNXGZBXFDNKOR-UHFFFAOYSA-N
SMILES:
COC1=CC=C(Br)C=C1F
1
–
1
of
1
results
1
4-Bromo-2-fluoroanisole 98.0+%, TCI America™
CAS: 2357-52-0 Molecular Formula: C7H6BrFO Molecular Weight (g/mol): 205.026 MDL Number: MFCD00011710 InChI Key: DWNXGZBXFDNKOR-UHFFFAOYSA-N Synonym: 4-bromo-2-fluoroanisole,2-fluoro-4-bromoanisole,1-bromo-3-fluoro-4-methoxybenzene,benzene, 4-bromo-2-fluoro-1-methoxy,2-fluoro-4-bromo anisol,2-fluoro-4-bromo anisole,4-bromo-2-fluoro-1-methoxy-benzene,4-bromo-2-fluorophenylmethyl ether,4-brom-2-fluor-1-methoxybenzol,pubchem1971 PubChem CID: 75378 IUPAC Name: 4-bromo-2-fluoro-1-methoxybenzene SMILES: COC1=C(C=C(C=C1)Br)F