accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 198206-33-6

FFFOI

CAS RN 198206-33-6

IUPAC Name: 1-iodo-3-(trifluoromethoxy)benzene
Synonyms: 1-iodo-3-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 288.01
Molecular Formula: C7H4F3IO
InChi Key: UQZXQSQWKJZHCD-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results