CAS RN 189028-93-1
CAS RN 189028-93-1
IUPAC Name:
1-(4-fluorophenyl)-5-(2-oxo-4-phenyl-1,3-oxazolidin-3-yl)pentane-1,5-dione
Synonyms:
1-(4-fluorophenyl)-5-(2-oxo-4-phenyl-1,3-oxazolidin-3-yl)pentane-1,5-dione
Molecular Weight (g/mol):
355.37
Molecular Formula:
C20H18FNO4
InChi Key:
XXSSRSVXDNUAQX-UHFFFAOYNA-N
SMILES:
FC1=CC=C(C=C1)C(=O)CCCC(=O)N1C(COC1=O)C1=CC=CC=C1
1
–
1
of
1
results
1
(S)-4-Phenyl-3-[5-(4-fluorophenyl)-5-oxopentanoyl]-2-oxazolidinone 98.0+%, TCI America™
CAS: 189028-93-1 Molecular Formula: C20H18FNO4 Molecular Weight (g/mol): 355.365 MDL Number: MFCD08061377 InChI Key: XXSSRSVXDNUAQX-QGZVFWFLSA-N PubChem CID: 11187377 IUPAC Name: 1-(4-fluorophenyl)-5-[(4S)-2-oxo-4-phenyl-1,3-oxazolidin-3-yl]pentane-1,5-dione SMILES: C1C(N(C(=O)O1)C(=O)CCCC(=O)C2=CC=C(C=C2)F)C3=CC=CC=C3