CAS RN 1888-94-4
CAS RN 1888-94-4
IUPAC Name:
2-chloroethyl 2-methylprop-2-enoate
Synonyms:
2-chloroethyl 2-methylprop-2-enoate
Molecular Weight (g/mol):
148.59
Molecular Formula:
C6H9ClO2
InChi Key:
GPOGMJLHWQHEGF-UHFFFAOYSA-N
SMILES:
CC(=C)C(=O)OCCCl
1
–
1
of
1
results
1
Thermo Scientific Chemicals 2-Chloroethyl methacrylate, 97%, stab. with 400-600ppm hydroquinone
CAS: 1888-94-4 Molecular Formula: C6H9ClO2 Molecular Weight (g/mol): 148.59 MDL Number: MFCD00045291 InChI Key: GPOGMJLHWQHEGF-UHFFFAOYSA-N Synonym: 2-chloroethyl methacrylate,chloroethyl methacrylate,methacrylic acid, 2-chloroethyl ester,2-propenoic acid, 2-methyl-, 2-chloroethyl ester,.beta.-chloroethyl methacrylate,methacrylic acid beta-chloroethyl ester,methacrylic acid .beta.-chloroethyl ester,acmc-20anxx,chloroethylmethacrylate,2-chloroethylmethacrylate PubChem CID: 15905 IUPAC Name: 2-chloroethyl 2-methylprop-2-enoate SMILES: CC(=C)C(=O)OCCCl