CAS RN 18013-97-3
CAS RN 18013-97-3
IUPAC Name:
1,4-diethyl 2,5-dibromobenzene-1,4-dicarboxylate
Synonyms:
1,4-diethyl 2,5-dibromobenzene-1,4-dicarboxylate
Molecular Weight (g/mol):
380.03
Molecular Formula:
C12H12Br2O4
InChi Key:
WXRSDHICEYICMV-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC(Br)=C(C=C1Br)C(=O)OCC
1
–
1
of
1
results
1
Diethyl 2,5-Dibromoterephthalate 98.0+%, TCI America™
CAS: 18013-97-3 Molecular Formula: C12H12Br2O4 Molecular Weight (g/mol): 380.032 MDL Number: MFCD12407163 InChI Key: WXRSDHICEYICMV-UHFFFAOYSA-N Synonym: 2,5-Dibromoterephthalic Acid Diethyl Ester PubChem CID: 22146229 IUPAC Name: diethyl 2,5-dibromobenzene-1,4-dicarboxylate SMILES: CCOC(=O)C1=CC(=C(C=C1Br)C(=O)OCC)Br