CAS RN 1781835-02-6
CAS RN 1781835-02-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ WAY 316606 hydrochloride
Secreted Frizzled-related protein-1 (sFRP-1) inhibitor; increases Wnt signaling