CAS RN 17766-28-8
CAS RN 17766-28-8
IUPAC Name:
1-cyclohexylpiperazine
Synonyms:
1-cyclohexylpiperazine
Molecular Weight (g/mol):
168.28
Molecular Formula:
C10H20N2
InChi Key:
XPDSXKIDJNKIQY-UHFFFAOYSA-N
SMILES:
C1CCC(CC1)N1CCNCC1
1
–
1
of
1
results
1
1-Cyclohexylpiperazine 98.0+%, TCI America™
CAS: 17766-28-8 Molecular Formula: C10H20N2 Molecular Weight (g/mol): 168.284 MDL Number: MFCD00044809 InChI Key: XPDSXKIDJNKIQY-UHFFFAOYSA-N Synonym: n-cyclohexylpiperazine,1-cyclohexyl-piperazine,cyclohexylpiperazine,piperazine, 1-cyclohexyl,1-cyclohexylpiperazin,n-cyclohexyl piperazine,1-cyclohexyl piperazine,piperazine,1-cyclohexyl,4-1-cyclohexyl piperazine,acmc-1c47v PubChem CID: 87298 IUPAC Name: 1-cyclohexylpiperazine SMILES: C1CCC(CC1)N2CCNCC2