accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17635-21-1

H3CNCH3CH3N

CAS RN 17635-21-1

IUPAC Name: 2,3,6-trimethylquinoxaline
Synonyms: 2,3,6-trimethylquinoxaline
Molecular Weight (g/mol): 172.23
Molecular Formula: C11H12N2
InChi Key: GQRWKGBOBWHKHP-UHFFFAOYSA-N
SMILES: CC1=CC=C2N=C(C)C(C)=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results